5-Methylindoline 95%
Catalog # APOSOR951911-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
- Environmentally Preferable:
Specifications
About this item
:
:133.1900 g/mol
Storage Temperature:Ambient
CAS Number:65826-95-1
Formula:C₉H₁₁N
MDL Number:MFCD03093636