Order Entry
Denmark
ContactUsLinkComponent
(1R)-(-)-Camphor-10-sulphonic acid 97%
(1R)-(-)-Camphor-10-sulphonic acid 97%
Catalog # APOSOR52399-250G
Supplier:  Apollo Scientific
CAS Number:  
(1R)-(-)-Camphor-10-sulphonic acid 97%
Catalog # APOSOR52399-250G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 g
  • Seq:
    3545
  • Environmentally Preferable:

Specifications

About this item

Synonyms: L(-)-Camphor-10-sulphonic acid, (1R)-(-)-2-Oxobornane-10-sulphonic acid, ((1R,4S)-7,7-Dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl)methanesulphonic acid, (-)-CSA
 :
 :232.3 g/mol
EINECS:252-817-9
UN:3261
 :GHS05
Storage Temperature:Ambient
CAS Number:35963-20-3
 :MFCD00064158
Formula:C₁₀H₁₆O₄S
MDL Number:MFCD00150753
Boiling Pt:375
Melting Pt:198 °C (dec.)
ADR:8 II