tert-Butyl-2,4-dioxopiperidine-1-carboxylate
Catalog # APOSOR52261-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
About this item
Synonyms: tert-Butyl 2,4-dioxopiperidine-1-carboxylate, Piperidine-2,4-dione, N-BOC protected
:
:213.2304 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:845267-78-9
Formula:C₁₀H₁₅NO₄
MDL Number:MFCD10566071
Melting Pt:111…112 °C