(2-Aminothiazol-4-yl)methanol
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:51307-43-8
:
Formula:C₄H₆N₂OS
MDL Number:MFCD08275702
Molecular Weight:130.17 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together