Isoquinoline-4-carbonitrile
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:34846-65-6
:
Formula:C₁₀H₆N₂
MDL Number:MFCD00957039
Molecular Weight:154.1700 g/mol
Melting Pt:104 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together