1-(Benzenesulphonyl)-4-chloro-1H-pyrrolo[2,3-b]pyridine 96%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 4-Chloro-1-benzenesulphonyl-7-azaindole, 4-Chloro-1-(phenylsulphonyl)-1H-pyrrolo[2,3-b]pyridine
CAS Number:744209-63-0
:
Formula:C₁₃H₉ClN₂O₂S
MDL Number:MFCD04153750
Molecular Weight:292.741 g/mol
Melting Pt:117…120 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together