(Triisopropylsilyl)acetylene
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: Ethynyltriisopropylsilane
CAS Number:89343-06-6
:
Formula:C₁₁H₂₂Si
MDL Number:MFCD00075452
Boiling Pt:50…52 °C/0.6mmHg
Density:0.813 g/cm³ (25 °C)
Molecular Weight:182.38 g/mol
Flash Pt:56 °C
Storage Temperature:Ambient
UN:1993
ADR:3 III
:GHS02
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together