0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Denmark
ContactUsLinkComponent
N-Acetyl-α-D-glucosamine
N-Acetyl-α-D-glucosamine
N-Acetyl-α-D-glucosamine
APOSOR28962-1G
:
Apollo Scientific
:
N-Acetyl-α-D-glucosamine
APOSOR28962-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
Environmentally Preferable:
:
2-Acetamido-2-deoxy-α-D-glucopyranose
:
:
233-115-1
:
GHS07
:
Ambient
:
10036-64-3
:
C₈H₁₅NO₆
:
211 °C