1-Amino-2-ethylbutane 95%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 2-Ethyl-1-butylamine, 2-Ethylbutan-1-amine, 2-Ethylbutylamine
CAS Number:617-79-8
:
Formula:C₆H₁₅N
MDL Number:MFCD00040755
Boiling Pt:125 °C
Density:0.776 g/cm³ (25 °C)
Molecular Weight:101.19 g/mol
Flash Pt:14 °C
EINECS:210-530-6
UN:3286
ADR:3+6.1+8 II
:GHS02
:GHS05
:GHS06
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together