5-Amino-2,3-dihydrobenzo[b]furan
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 2,3-Dihydrobenzofuran-5-amine, 5-Amino-2,3-dihydrobenzofuran, 2,3-dihydro-1-benzofuran-5-amine, 5-Benzofuranamine,2,3-dihydro-
CAS Number:42933-43-7
:
Formula:C₈H₉NO
MDL Number:MFCD03617969
Molecular Weight:135.166 g/mol
Melting Pt:78…83 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together