1-Bromo-4-octylbenzene
Catalog # APOSOR21923-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
About this item
Synonyms: (4-Bromophenyl)octane
:
:269.22 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:51554-93-9
Formula:C₁₄H₂₁Br
MDL Number:MFCD00038348