Vitamin B12 (cyanocobalamin) USP
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:68-19-9
:
Formula:C₆₃H₈₈CoN₁₄O₁₄P
MDL Number:MFCD00151092
Molecular Weight:1355.38 g/mol
Melting Pt:>300 °C
Merck Index:12,10152
:GHS07
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together