4-Chloro-2-ethoxy-5-fluoropyrimidine 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:56076-20-1
:
Formula:C₆H₆ClFN₂O
MDL Number:MFCD06410760
Molecular Weight:176.576 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together