3,5-Difluoro-4-methoxyphenylacetonitrile 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:886498-79-9
:
Formula:C₉H₇F₂NO
MDL Number:MFCD04115920
Molecular Weight:183.155 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together