5-(Piperidin-1-yl)pyridin-2-amine 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:94924-94-4
:
Formula:C₁₀H₁₅N₃
MDL Number:MFCD10009548
Molecular Weight:177.246 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together