Order Entry
Denmark
ContactUsLinkComponent
2,2'-((2-((2-Hydroxyethyl)amino)ethyl)azanediyl)bis(ethan-1-ol) 95%
2,2'-((2-((2-Hydroxyethyl)amino)ethyl)azanediyl)bis(ethan-1-ol) 95%
Catalog # APOSOR321430-250MG
Supplier:  Apollo Scientific
CAS Number:  
2,2'-((2-((2-Hydroxyethyl)amino)ethyl)azanediyl)bis(ethan-1-ol) 95%
Catalog # APOSOR321430-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 2,2'-((2-((2-Hydroxyethyl)amino)ethyl)azanediyl)diethanol
 :
 :192.256 g/mol
Storage Temperature:Ambient
CAS Number:60487-26-5
Formula:C₈H₂₀N₂O₃
MDL Number:MFCD20667837