4-Benzyl-2-(hydroxymethyl)piperazine 95+%
Catalog # APOSOR13971-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
- Environmentally Preferable:
Specifications
About this item
Synonyms: (4-Benzylpiperazin-2-yl)methanol
:
:206.284 g/mol
Storage Temperature:Ambient
CAS Number:85817-34-1
Formula:C₁₂H₁₈N₂O
MDL Number:MFCD03001699