5-(Ethoxycarbonyl)thiophene-3-boronic acid
Catalog # APOSOR11699-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
About this item
Synonyms: (5-(Ethoxycarbonyl)thiophen-3-yl)boronic acid, Ethyl 4-boronothiophene-2-carboxylate, 4-Borono-2-(ethoxycarbonyl)thiophene
:
:200.02 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:957121-19-6
Formula:C₇H₉BO₄S
MDL Number:MFCD09800891