Acid Fuchsin calcium salt ≥60 % (dye content), certified by the Biological Stain Commission, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
Synonyms: Rubin S, Fuchsin S, SS, SS, ST, or SIII, Acid Rubin, Acid Magenta
:
Formula:C₂H₁₇CaN₃O₉S₃
MDL Number:MFCD00001654
Molecular Weight:579.64 g/mol
:M-Clarity™ quality level = MQ200
Storage Temperature:Room Temperature
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance (Form)
Powder or Crystalline Powder:Conforms to Requirements - Infrared spectrum:Conforms to Structure
- Wavelength
C=0.01g/l in H2O + 2.5ml 0.1N HCl:541 - 548 nm - Extinction Coefficient:≥50000
- Wavelength :285 - 291 nm
- Biological Stain Commission
Commission Certified Number::Pass - Dye Content:≥ 60%
- Carbon:≥ 28.5%
- Nitrogen:≥5.0%
- Calcium (Ca)
% Calcium by ICP Assay:≥4.7 %
Specification Test Results
Related Information
Frequently Bought Together