Adrenochrome, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
Synonyms: 3-Hydroxy-1-methyl-5,6-indolinedione
CAS Number:54-06-8
:
Formula:C₉H₉NO₃
Molecular Weight:179.175 g/mol
:M-Clarity™ quality level = MQ200
Storage Temperature:Freezer
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance (Colour):Red to Brown
- Appearance (Form):Powder
- Infrared spectrum:Conforms to Structure
- EmM
At 299 to 304 nm in Water:9.0 - 10.5 - Wavelength:299 - 304 nm
- EmM
At 485 to 490 nm in Water:3.0 - 4.5 - Carbon:57.0 - 63.8 %
- Nitrogen:7.4 - 8.3 %
- Purity (HPLC):≥ 95 %
Specification Test Results
Frequently Bought Together