Bottles, UN approved, with self-sealing screw cap
Sturdy HDPE bottles are supplied with tamper-evident screw caps.
- Robust HDPE bottles
- Good chemical resistance
- UN approved for liquids
- Leakproof design
Bottles are ideal for sample shipping.
Frequently Bought Together