1,3,5-Tris[4-(diphenylamino)phenyl]benzene ≥95.0% (by total nitrogen basis)
Supplier: TCI
Certificates
About this item
CAS Number:147951-36-8
:
Formula:C₆₀H₄₅N₃
MDL Number:MFCD30721946
Molecular Weight:808.04 g/mol
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 95.0 area%
- Elemental analysis (Nitrogen):4.80 to 5.50 %
Specification Test Results
Frequently Bought Together