GMBS (N-Succinimidyl 4-maleimidobutyrate) ≥98.0% (by HPLC, total nitrogen) cross-linking reagent
Supplier: TCI
Certificates
About this item
Synonyms: N-(γ-Maleimidobutyryloxy)succinimide, 2,5-Dioxopyrrolidin-1-yl 4-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)butanoate, N-(4-Maleimidobutyryloxy)succinimide, GMBS, 4-Maleimidobutyric acid N-hydroxysuccinimide ester, 4-Maleimidobutyric acid N-succinimidyl ester
CAS Number:80307-12-6
:
Formula:C₁₂H₁₂N₂O₆
MDL Number:MFCD00036817
Molecular Weight:280.24 g/mol
:GHS07
Storage Temperature:Freezer
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light red powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:126.0 to 131.0 deg-C
Specification Test Results
Frequently Bought Together