Resiquimod ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
Synonyms: 1-[4-Amino-2-(ethoxymethyl)-1H-imidazo[4,5-c]quinolin-1-yl]-2-methylpropan-2-ol, 4-Amino-2-(ethoxymethyl)-a,a-dimethyl-1H-imidazo[4,5-c]quinoline-1-ethanol, 1-(4-Amino-2-(ethoxymethyl)-1H-imidazo[4,5-c]quinolin-1-yl)-2-methylpropan-2-ol, 4-Amino-2-(ethoxymethyl)-alpha,alpha-dimethyl-1H-imidazo[4,5-c]quinoline-1-ethanol, R-848
CAS Number:144875-48-9
:
Formula:C₁₇H₂₂N₄O₂
MDL Number:MFCD00937759
Molecular Weight:314.389 g/mol
Melting Pt:193°C
Merck Index:14,08151
Storage Temperature:Freezer
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Elemental analysis (Nitrogen):17.00 to 18.50 %
Specification Test Results
Frequently Bought Together