PA-824 ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
Synonyms: (S)-2-Nitro-6-[[4-(trifluoromethoxy)benzyl]oxy]-6,7-dihydro-5H-imidazo[2,1-b][1,3]oxazine, Pretomanid, (S)-2-Nitro-6-((4-(trifluoromethoxy)benzyl)oxy)-6,7-dihydro-5H-imidazo[2,1-b][1,3]oxazine
CAS Number:187235-37-6
:
Formula:C₁₄H₁₂F₃N₃O₅
MDL Number:MFCD06809939
Molecular Weight:359.26 g/mol
Storage Temperature:Freezer
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:148.0 to 152.0 deg-C
- Specific rotation [a]20/D:-44.0 to -49.0 deg (C=1, MeOH)
- Elemental analysis (Nitrogen):11.40 to 12.10 %
Specification Test Results
Frequently Bought Together