MES sodium salt ≥85.0% for biopharmaceutical production, purified, J.T.Baker®
Supplier: Avantor
Certificates
About this item
Synonyms: Sodium 2-Morpholinoethanesulphonate, Sodium 3-morpholin-1-ylethylsulphonate, 2-(N-Morpholino)ethanesulphonic acid sodium salt, Sodium 2-morpholinoethane-1-sulphonate, MES-Na
CAS Number:71119-23-8
:438589-24-3
Formula:C₆H₁₂NNaO₄S
MDL Number:MFCD00065473
Molecular Weight:217.22 g/mol
EINECS:275-203-2
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:≥85.0%
- Assay (C₆H₁₂NNaO₄S):≥85.0 %
- Appearance:Passes Test
- Identification (by IR):Passes Test
- pH of 1% Aqueous Solution at 25°C:8.9 - 10.1
- Solubility:Passes Test
- Ultraviolet Absorbance of a 10% Aqueous Solution - 280 nm:≤0.03
- Ultraviolet Absorbance of a 10% Aqueous Solution - 260 nm:≤0.05
- Water (H₂O):≤15.0 %
Specification Test Results
Frequently Bought Together