5,12-Naphthacenequinone ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
Synonyms: 2,3-Benzo-9,10-anthraquinone, Tetracenequinone
CAS Number:1090-13-7
:
Formula:C₁₈H₁₀O₂
MDL Number:MFCD00003701
Molecular Weight:258.28 g/mol
EINECS:214-127-6
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Yellow to Orange powder to crystal
- Purity (GC):min. 98.0 %
- Solubility in hot DMF:almost transparency
Specification Test Results
Frequently Bought Together