2-(2-Methoxyphenyl)ethanol ≥98.0% (by GC)
: TCI
:7417-18-7
:MFCD00002889
:C₉H₁₂O₂
:134°C
:152.19 g/mol
:231-029-9
- Environmentally Preferable:
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
Frequently Bought Together