Order Entry
Denmark
ContactUsLinkComponent
4-(4-Methyl-1,3-dioxolan-2-yl)-1,2-methylenedioxybenzene ≥98.0% (by GC)
4-(4-Methyl-1,3-dioxolan-2-yl)-1,2-methylenedioxybenzene ≥98.0% (by GC)
  TCIAM2483-25G
 :  TCI
 :  
undefined
4-(4-Methyl-1,3-dioxolan-2-yl)-1,2-methylenedioxybenzene ≥98.0% (by GC)
  TCIAM2483-25G
 :  TCI
 :  

 

  • Pk:
    25 g
  • Seq:
    3308
  • Environmentally Preferable:

 

 

  • Appearance:
    Colorless to Almost colorless clear liquid to slightly cloudy liquid
  • Purity (GC):
    min. 98.0 %

 

 

 : 5-(4-Methyl-1,3-dioxolan-2-yl)benzo[d][1,3]dioxole, 5-(4-Methyl-1,3-dioxolan-2-yl)-1,3-benzodioxole, Piperonal Propylene Glycol Acetal
 :
 :208.21 g/mol
 :262-897-7
 :GHS07
 :61683-99-6
 :C₁₁H₁₂O₄
 :MFCD13194964