Order Entry
Denmark
ContactUsLinkComponent
Methyl trans-p-coumarate ≥98.0% (by GC)
Methyl trans-p-coumarate ≥98.0% (by GC)
Supplier:  TCI
Certificates
undefined
Methyl trans-p-coumarate ≥98.0% (by GC)
Supplier:  TCI

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    137.0 to 141.0 deg-C

Specification Test Results

Related Information

Frequently Bought Together