Order Entry
Denmark
ContactUsLinkComponent
4-Isopropyl-4'-methyldiphenyliodonium tetrakis(pentafluorophenyl)borate ≥98.0% (by titrimetric analysis)
4-Isopropyl-4'-methyldiphenyliodonium tetrakis(pentafluorophenyl)borate ≥98.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
4-Isopropyl-4'-methyldiphenyliodonium tetrakis(pentafluorophenyl)borate ≥98.0% (by titrimetric analysis)
Supplier:  TCI

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Ion exchange titration):
    min. 98.0 %
  • Melting point:
    131.0 to 135.0 deg-C

Specification Test Results

Related Information

Frequently Bought Together