(R)-1-(pyridin-4-yl)ethanol ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
Synonyms: (R)-alpha-Methyl-4-pyridinemethanol, (R)-1-(4-Pyridyl)ethanol, (R)-4-(1-Hydroxyethyl)pyridine, (R)-1-(Pyridin-4-yl)ethanol
CAS Number:27854-88-2
:
Formula:C₇H₉NO
MDL Number:MFCD00077865
Molecular Weight:123.155 g/mol
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Light red to Green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 96.0 %
- Optical purity (GC):min. 98.0 ee%
- Melting point:67.0 to 71.0 deg-C
Specification Test Results
Frequently Bought Together