Order Entry
Denmark
ContactUsLinkComponent
4-Perhydroazepinone hydrochloride ≥98.0% (by titrimetric analysis)
4-Perhydroazepinone hydrochloride ≥98.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
4-Perhydroazepinone hydrochloride ≥98.0% (by titrimetric analysis)
Supplier:  TCI

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (Argentmetric Titration):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 97.0 %
  • Melting point:
    168.0 to 172.0 deg-C

Specification Test Results

Frequently Bought Together