2-Hydroxyanthracene ≥93.0% (by GC)
Supplier: TCI
Certificates
About this item
Synonyms: Anthracen-2-ol
CAS Number:613-14-9
:
Formula:C₁₄H₁₀O
MDL Number:MFCD06653605
Molecular Weight:194.23 g/mol
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Light yellow to Dark green powder to crystaline
- Purity (GC):min. 93.0 %
Specification Test Results
Frequently Bought Together