Order Entry
Denmark
ContactUsLinkComponent
1-Ethyl-2-methylcyclohexane ≥98.0% (by GC)
1-Ethyl-2-methylcyclohexane ≥98.0% (by GC)
Catalog # TCIAE0422-1ML
Supplier:  TCI
CAS Number:  
1-Ethyl-2-methylcyclohexane ≥98.0% (by GC)
Catalog # TCIAE0422-1ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Seq:
    2070
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :126.24 g/mol
UN:3295
 :GHS02
CAS Number:3728-54-9
Formula:C₉H₁₈
MDL Number:MFCD00067158
Boiling Pt:156°C
ADR:3 III