1,5-Dibromo-2,4-dimethylbenzene ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
Synonyms: 4,6-Dibromo-m-xylene
CAS Number:615-87-2
:
Formula:C₈H₈Br₂
MDL Number:MFCD00154984
Boiling Pt:256°C
Molecular Weight:263.96 g/mol
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:69.0 to 73.0 deg-C
Specification Test Results
Frequently Bought Together