Order Entry
Denmark
ContactUsLinkComponent
Dibutyryl-cAMP sodium salt ≥97.0% (by HPLC)
Dibutyryl-cAMP sodium salt ≥97.0% (by HPLC)
Catalog # TCIAD4228-25MG
Supplier:  TCI
CAS Number:  
Dibutyryl-cAMP sodium salt ≥97.0% (by HPLC)
Catalog # TCIAD4228-25MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Seq:
    2315
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 97.0 area%
  • Elemental analysis (Nitrogen):
    13.40 to 14.60 %

Specification Test Results

About this item

Synonyms: Bucladesine sodium salt, DC-2797, Dibutyl cyclic AMP sodium salt, Dibutyryl-cAMP, N6,2'-O-Dibutyryladenosine 3',5'-Cyclic monophosphate sodium salt, DbcAMP, Sodium (4aR,6R,7R,7aR)-6-(6-butyramido-9H-purin-9-yl)-7-(butyryloxy)tetrahydro-4H-furo[3,2-d][1,3,2]dioxaphosphinin-2-olate 2-oxide, Bucladesine
 :
 :491.37 g/mol
Storage Temperature:Freezer
EINECS:241-059-4
CAS Number:16980-89-5
Formula:C₁₈H₂₃N₅O₈P·Na
MDL Number:MFCD00005843
Merck Index:14,01463