4-(10-Bromoanthracen-9-yl)benzonitrile ≥98.0% (by GC)
Catalog # TCIAB6114-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:200.0 to 204.0 deg-C
Specification Test Results
About this item
Synonyms: 4-(10-Bromo-9-anthryl)benzonitrile, 4-(10-Bromo-9-anthracenyl)benzonitrile
:
:358.24 g/mol
:GHS07
CAS Number:937372-45-7
Formula:C₂₁H₁₂BrN
MDL Number:MFCD30730126