2-Benzyloxy-5-pyridineboronic acid (contains varying amounts of Anhydride)
Supplier: TCI
Certificates
About this item
Synonyms: 6-Benzyloxypyridine-3-boronic acid, 6-(Benzyloxy)pyridine-3-boronic Acid
CAS Number:929250-35-1
:
Formula:C₁₂H₁₂BNO₃
MDL Number:MFCD06798262
Molecular Weight:229.04 g/mol
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):98.0 to 108.0 %
Specification Test Results
Frequently Bought Together