2-(2H-Benzotriazol-2-yl)-4-methyl-6-(2-propenyl)phenol ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
Synonyms: 2-Allyl-6-(2H-benzotriazol-2-yl)-4-methylphenol
CAS Number:2170-39-0
:
Formula:C₁₆H₁₅N₃O
MDL Number:MFCD00236141
Molecular Weight:265.32 g/mol
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Purity (GC):min. 98.0 %
- Melting point:98.0 to 102.0 deg-C
Specification Test Results
Frequently Bought Together