2-Methoxy-1-methylethyl acetate (PGMEA) 99%
: Propylene glycol 1-methyl ether 2-acetat, 2-(1-Methoxy)propyl acetate, 1-Methoxy-2-propyl acetate, Propylene glycol methyl ether acetat, 1,2-Propanediol monomethyl ether acetate, DOWANOL PMA
:108-65-6
:
:C₆H₁₂O₃
:MFCD00038500
:145.8 °C (1013 hPa)
:–87 °C
:0.96 g/cm³
:132.16 g/mol
:42 °C
:Ambient
:203-603-9
:1993
:3 III
:GHS02
- Environmentally Preferable:
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4010 to 1.4030 (20°C, 589 nm)
- Water:≤0.05 %
- GC:≥98.5 %
- Acidity (CH3COOH):≤0.02 %
Frequently Bought Together