2-Bromo-6-methoxyphenol ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:28165-49-3
:
Formula:C₇H₇BrO₂
MDL Number:MFCD08146628
Boiling Pt:146°C
Molecular Weight:203.04 g/mol
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:62.0 to 66.0 deg-C
Specification Test Results
Frequently Bought Together