5-Bromo-2-hydroxy-4-picoline ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
Synonyms: 5-Bromo-2-hydroxy-4-methylpyridine
CAS Number:164513-38-6
:
Formula:C₆H₆BrNO
MDL Number:MFCD03427664
Molecular Weight:188.02 g/mol
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:199.0 to 203.0 deg-C
Specification Test Results
Frequently Bought Together