5-Bromo-3-indolyl-β-D-galactopyranoside ≥98.0% (by HPLC) for biochemical research
Supplier: TCI
Certificates
About this item
Synonyms: Bluo-Gal, (2S,3R,4S,5R,6R)-2-((5-Bromo-1H-indol-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol, 5-Bromo-3-indolyl β-D-galactopyranoside
CAS Number:97753-82-7
:
Formula:C₁₄H₁₆BrNO₆
MDL Number:MFCD00063691
Molecular Weight:374.19 g/mol
Storage Temperature:Freezer
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- beta-Galactosidase detection test:to pass test
Specification Test Results
Frequently Bought Together