Acrivastine ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
Synonyms: (E)-3-(6-((E)-3-(Pyrrolidin-1-yl)-1-(p-tolyl)prop-1-en-1-yl)pyridin-2-yl)acrylic acid
CAS Number:87848-99-5
:
Formula:C₂₂H₂₄N₂O₂
MDL Number:MFCD00869830
Molecular Weight:348.45 g/mol
Merck Index:14,00127
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Elemental analysis (Nitrogen):7.50 to 8.50 %
Specification Test Results
Frequently Bought Together