2-Anilino-9,9-dimethylfluorene ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
Synonyms: 9,9-Dimethyl-N-phenyl-9H-fluoren-2-amine
CAS Number:355832-04-1
:
Formula:C₂₁H₁₉N
MDL Number:MFCD20040462
Molecular Weight:285.39 g/mol
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:99.0 to 103.0 deg-C
Specification Test Results
Frequently Bought Together