Order Entry
Denmark
ContactUsLinkComponent
 
L(+)-Calcium lactate pentahydrate 98%
Catalog # ACRO250200010
CAS Number:  
undefined
L(+)-Calcium lactate pentahydrate 98%
Catalog # ACRO250200010
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 kg
  • Seq:
    4024
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    White Powder
  • Infrared spectrum:
    Conforms
  • Titration Complexometric:
    ≥97.5 % (on dry substance)
  • Loss on drying:
    22.0 % to 27.0 %
  • Heavy metals:
    ≤10 ppm
  • pH:
    6.0 to 8.0 (5 % in water)
  • Chloride (Cl):
    ≤80 ppm
  • Phosphate (PO4):
    ≤150 ppm
  • Sulfate (SO4):
    ≤400 ppm
  • Arsenic (As):
    ≤1 ppm
  • Calcium (Ca):
    13.4 to 14.5 %
  • Iron (Fe):
    ≤40 ppm
  • Alkalies+Mg (as sulfates):
    ≤1.0 %

Specification Test Results

About this item

Synonyms: L(+)-Lactic acid calcium salt pentahydrate, Calcium (S)-(+)-2-hydroxypropionate pentahydrate, (S)-(+)-2-Hydroxypropionic acid calcium salt pentahydrate, (S)-(+)-Lactic acid calcium salt pentahydrate, (S)-(+)-2-Hydroxypropanoic acid calcium salt pentahydrate, Calcium L(+)-2-hydroxypropionate pentahydrate
 :
 :308.3 g/mol
Storage Temperature:Ambient
EINECS:248-953-3
CAS Number:865717-20-0
 :5743-47-5
Formula:C₆H₁₀CaO₆·5H₂O
MDL Number:MFCD00150344
Melting Pt:240