Order Entry
Denmark
ContactUsLinkComponent
 
D-(-)-Glucamine ≥97.0% (by titrimetric analysis)
 :  TCI
 
undefined
D-(-)-Glucamine ≥97.0% (by titrimetric analysis)
 :  TCI

 

  • Environmentally Preferable:

 

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Neutralization titration):
    min. 97.0 %
  • Specific rotation [a]20/D:
    -5.0 to -8.0 deg (C=10, H2O)

 

Frequently Bought Together