Anti-SARS-CoV-2 Nucleocapsid Protein Human Monoclonal Antibody [clone: HC2003]
Catalog # GENSA02039-100
Supplier: Genscript
Specifications
- Antibody type:Primary
- Antigen name:SARS-CoV-2 Nucleocapsid Protein
- Clonality:Monoclonal
- Clone:HC2003
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Human
- Isotype:IgG1
- Reactivity:Human
- Environmentally Preferable:
- Epitope:Confidentiality
- Cross adsorption:No
- Format:Purified antibody
- Form:liquid
- Gene ID:MN908947
- Antigen synonyms:N Protein/NP
- Amino acid number:Confidentiality
- Modification:Humanization
- Antibody fragment:Whole Molecule
- Storage buffer:PBS, pH 7.4, containing 0.02% sodium azide.
- Molecular weight:150 kDa
- Sequence:Confidentiality
- Storage temperature:20° C
- Concentration:1 mg/ml, supplied in PBS, pH 7.4, containing 0.02% sodium azide.
- Shipping temperature:4° C
- Immunogen:Confidentiality
- Purification:Protein A
- Pk:100 µG
Specifications
About this item
SARS-CoV-2 (Severe acute respiratory syndrome coronavirus 2), also known as 2019-nCoV, is a positive-sense single-stranded RNA virus. It caused coronavirus disease 2019 (COVID-19). Nucleocapsid Protein is a most abundant structure protein of the coronavirus which is associated with nucleic acid.
SARS-CoV-2 Nucleocapsid Antibody (HC2003), Human Chimeric is produced from cell culture in vitro under conditions free from animal-derived components.
Type: Primary
Antigen: Protein
Clonality: Monoclonal
Clone: HC2003 (monoclonal antibody)
Conjugation: Unconjugated
Epitope: Confidentiality
Host: Human
Isotype: Recombinant human IgG1
Reactivity: Human