Order Entry
Denmark
ContactUsLinkComponent
Nitric acid solution ~32,5%, special quality
Nitric acid solution ~32,5%, special quality
Catalog # HONC30772H-2.5L
CAS Number:  
Nitric acid solution ~32,5%, special quality
Catalog # HONC30772H-2.5L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay:
    32.2 - 32.8 %

Specification Test Results

About this item

Synonyms: Nitric acid solution
 :
 :63.01 g/mol
UN:2031
 :GHS05
 :GHS06
CAS Number:7697-37-2
Formula:HNO₃
MDL Number:MFCD00011349
Boiling Pt:100 °C
Melting Pt:-42
Density:1.196 g/cm3
ADR:8 II
Merck Index:13,06610